Tissue embedding sponge, biopsy test
Artikelnr. 631-9681
Lieferant: DELTALAB
Spezifikationen
- Colour:Blue
- Description:Biopsy sponge, foam
- Dimensions:31×25×2,2 mm
- Pk:500
Spezifikationen
Über diesen Artikel
Foam sponge, very useful for small specimens in biopsy tests.
- Avoids sample loss though drain holes when sample is processed
- Does not affect parafin drainage
Fits standard cassettes.