Racks for upright freezers, with sliding shelves and depth 670 mm, Comfort racks
: TENAK
Stainless steel.
- Racks for four different box heights are available
- Fit all standard cryoboxes
For upright freezers. These racks ensure a more ergonomic operating process. The cryoboxes are placed in drawers in the sliding shelf rack, increasing flexibility.
Note: Measure the inside height and depth of your freezer to ensure the right choice of rack.
: Delivered without cryoboxes.
Frequently Bought Together