ContactUsLinkComponent
Staining jar acc.to Coplin
Staining jar acc.to Coplin
Catalog # 631-9730
Supplier:  DELTALAB
Staining jar acc.to Coplin
Catalog # 631-9730
Supplier:  DELTALAB
Specifications
  • Ø:
    60 mm
  • Description:
    Staining jar, Coplin
  • Height:
    110 mm
  • Pk:
    1
Specifications
About this item
Jar made of PP. Rectangular container with rounded base and screw ribbed flat cap with an inner ring for preventing leakage.
  • Unbreakable
  • Ideal for used as a storage system
  • Good chemical and temperature resistance
These staining jar hold up to 10 slides (Two per groove).