Order Entry
Austria
ContactUsLinkComponent
Iodide Iodate concentrate for dilution to c(I₂) = 1/128 mol/l (1/64 N) in aqueous solution, Titrisol® volumetric solution, Supelco®
Iodide Iodate concentrate for dilution to c(I₂) = 1/128 mol/l (1/64 N) in aqueous solution, Titrisol® volumetric solution, Supelco®
Catalog # 1.09914.0001
Supplier:  Merck
CAS Number:  
undefined
Iodide Iodate concentrate for dilution to c(I₂) = 1/128 mol/l (1/64 N) in aqueous solution, Titrisol® volumetric solution, Supelco®
Catalog # 1.09914.0001
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1 Ampoul
  • Seq:
    0501

Specifications

Specification Test Results

  • Solubility:
    (20 °C) soluble
  • Density:
    1.07 g/cm3 (20 °C)
  • pH:
    13 (H2O, 20 °C)

Specification Test Results

About this item

 :
 :M-Clarity™ quality level = MQ100
 :GHS08
 :GHS05
Storage Temperature:Ambient