Orders LinkContactUsLinkComponent
Autoclavable baskets
Autoclavable baskets
Autoclavable baskets
Supplier:  DELTALAB
Certificates
Autoclavable baskets
Autoclavable baskets
3 Options Available Below
Product Details & Documents
Made of stainless steel wire.
  • Stackable
  • Good chemical resistance
  • Excellent temperature resistance
Baskets for autoclaves and carrying.
Documents
Product Family Options
Product Information
  • W×D×H
    Availability
    Price
  • 470×300×200 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 361,00 €
    361,00 €
    1 * 1 ST
       361,00 €
  • 570×300×150 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 197,00 €
    197,00 €
    1 * 1 ST
       197,00 €
  • 570×300×200 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 257,00 €
    257,00 €
    1 * 1 ST
       257,00 €