Order Entry
Austria
Orders LinkContactUsLinkComponent
Phenol red solution indicator, Supelco®

Phenol red solution indicator, Supelco®

Glass bottle, 100 mL
Catalog # 1.07242.0100
Supplier:   Merck
Phenol red solution indicator, Supelco®
Catalog # 1.07242.0100
Supplier:  Merck
CAS Number:  143-74-8
Phenol red solution indicator, Supelco®

Glass bottle, 100 mL

CAS Number:  143-74-8
Price
83,90 €
Log in for availability
1 * 100 mL
Product Details & Documents
Synonyms:  3,3-Bis(4-hydroxyphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide
  • CAS Number:143-74-8
  • Formula:C₁₉H₁₄O₅S
  • MDL Number:MFCD00003552
  • Molecular Weight:354.38 g/mol
  • Density:0.99 g/cm3 (20 °C)
  •  :M-Clarity™ quality level = MQ200
  • Storage Temperature:Ambient

Documents

Specifications

Specification Test Results

  • Application test:
    conforms
  • pH-value (20 °C):
    7.0 - 7.3

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL

Specifications