Order Entry
Austria
ContactUsLinkComponent
pH Indicator solution '4.5' acc. to Mortimer, Supelco®
pH Indicator solution '4.5' acc. to Mortimer, Supelco®
Catalog # 1.01359.0250
Supplier:  Merck
CAS Number:  
pH Indicator solution '4.5' acc. to Mortimer, Supelco®
Catalog # 1.01359.0250
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    250 mL
  • Seq:
    2590

Specifications

Specification Test Results

  • Application test:
    conforms
  • pH-value (20 °C):
    4.5 - 5.0

Specification Test Results

About this item

 :
Density:0.93 g/cm3 (20 °C)
Flash Pt:9 °C
Storage Temperature:Ambient
ADR:3,II
 :M-Clarity™ quality level = MQ200
UN:1170
 :GHS02