ContactUsLinkComponent
Drum tap, zinc
Drum tap, zinc
Catalog # MSGA2796
Supplier:  asecos
undefined
Drum tap, zinc
Catalog # MSGA2796
Supplier:  asecos
Specifications
  • Thread:
    3/4"
  • Description:
    Self-closing with flame guard and PTFE seal, fine dosing
  • Pk:
    1
Specifications
About this item
This fine dosing tap is made of zinc injection moulding.
  • Self closing
  • Flame guard
  • PTFE seal