Orders LinkContactUsLinkComponent
Drum taps, brass | Avantor
Drum taps, brass
Supplier:   asecos
Drum taps, brass
Supplier:  asecos
Certificates
Drum taps, brass
Drum taps, brass
3 Options Available Below
Product Details & Documents
Brass taps for drums with screw thread.
  • Self closing
  • Flame guard
  • PTFE seal
Ordering information: MSGA2799 includes a trailer fitting for decanting vessels.
Product Family Options
Product Information
  • For
    Thread
    Availability
    Price
  • MSGA2799
    Self-closing with flame guard, PTFE seal and trailer fitting
    Self-closing with flame guard, PTFE seal and trailer fitting
    Non-corrosive liquids <30 SAE
    3/4"
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 71,30 €
    71,30 €
    1 * 1 ST
       71,30 €
  • MSGA2798
    Self-closing with flame guard and PTFE seal
    Self-closing with flame guard and PTFE seal
    Aggressive liquids
    3/4"
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 68,20 €
    68,20 €
    1 * 1 ST
       68,20 €
  • MSGA2800
    Self-closing with flame guard and PTFE seal
    Self-closing with flame guard and PTFE seal
    Viscous liquids >30 SAE
    2”
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 202,00 €
    202,00 €
    1 * 1 ST
       202,00 €
Empfehlungen
Empfehlungen werden nur dann basierend auf Ihren Einkaufspräferenzen personalisiert, wenn Sie auf der Seite "Persönliche Daten" die Option „Mein Einkaufserlebnis verbessern“ aktiviert haben.
Andernfalls erhalten Sie allgemeine Empfehlungen.