Order Entry
Austria
ContactUsLinkComponent
Soil sieves
Soil sieves
Catalog # NICK9130
Supplier:  NICKEL ELECTRO
undefined
Soil sieves
Catalog # NICK9130
Supplier:  NICKEL ELECTRO

Specifications

  • Ø:
    100 mm
  • Mesh size:
    2,00 / 0,900 / 0,600 / 0,400 / 0,250 / 0,200 / 0,140 / 0,125 / 0,063 mm
  • Height:
    45 mm
  • Seq:
    01
  • Pk:
    1

Specifications

About this item

These soil sieves designed for educational use have an aluminium body with stainless steel mesh.

This complete set of soil sieves consists of nine sieve sizes with a lid and pan.