ContactUsLinkComponent
Pipette controller, Pipet-Aid® Hood Mate®
Pipette controller, Pipet-Aid® Hood Mate®
Catalog # DRUM4-000-302
Supplier:  DRUMMOND
Pipette controller, Pipet-Aid® Hood Mate®
Catalog # DRUM4-000-302
Supplier:  DRUMMOND
Specifications
  • Capacity:
    1 - 100 ml
  • Description:
    Pipette controller, Pipet-Aid® Hood Mate®, with EU-plug
  • Pk:
    1
Specifications
About this item
This controller is specifically designed for convenient, comfortable pipetting under a hood. Ideal for transfering liquids with the use of serological, volumetric and/or blow-out pipettes in either glass, plastic or something similar.
  • Longer coiled hose
  • Lightweight and manufactured from UV resistant materials
  • Extremely lightweight handle provides nine fill and empty speed settings and proportional fingertip control
  • Handle holder incorporates 'auto-off' feature: automatically shuts unit off after 3 m of non-use
  • Dedicated pump housing attaches to hood wall which saves space and prevents spills
  • Coiled vacuum/pressure line entering handle from the top prevents accidental knocking over of bench items