Order Entry
Austria
ContactUsLinkComponent
Cooling boxes, FlipLid™ 6
Cooling boxes, FlipLid™ 6
Catalog # CAMP2000036076
Supplier:  CAMPING GAZ
Cooling boxes, FlipLid™ 6
Catalog # CAMP2000036076
Supplier:  CAMPING GAZ

Specifications

  • Capacity:
    4,7 l
  • Colour:
    Blue
  • Description:
    Holds 6×0,33 l cans, weight: 0,7 kg
  • Dimensions:
    270×200×180 mm (ext); 220×160×130 mm (int)
  • Pk:
    1

Specifications

About this item

Personal cooler, insulated with polyurethane, the outer case and inner liner is made of HDPE material.

  • With ergonomic handle
  • Built-in locking system
  • Lid doubles as a tray