Order Entry
Austria
ContactUsLinkComponent
 
Ethidium bromide stock solution, Biotechnology Grade
Catalog # X328-10ML
Supplier:  VWR Chemicals
CAS Number:  
undefined
Ethidium bromide stock solution, Biotechnology Grade
Catalog # X328-10ML
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    10 mL
  • Seq:
    2275

Specifications

Specification Test Results

  • DNase:
    none detected
  • Moisture:
    <= 8%
  • Purity (Dry Basis):
    > = 98%
  • RNase:
    none detected

Specification Test Results

About this item

 :
 :394.31 g/mol
Storage Temperature:Ambient
CAS Number:1239-45-8
Formula:C₂₁H₂₀BrN₃
MDL Number:MFCD00011724