ContactUsLinkComponent
Weighing paper, grade MN 226
Weighing paper, grade MN 226
Catalog # 611-9091
Supplier:  MACHEREY-NAGEL
undefined
Weighing paper, grade MN 226
Catalog # 611-9091
Supplier:  MACHEREY-NAGEL
Specifications
  • Size:
    90×115 mm
  • Pk:
    100
  • Weight:
    40 g/m²
Specifications
About this item
This is a transparent paper, smooth on both sides, which can be used as substitute for weighing boats. The smooth surface of the paper guarantees that the weighed goods can be transferred without loss.