Order Entry
Austria
ContactUsLinkComponent
E.Z.N.A.® Endo-Free plasmid kits
E.Z.N.A.® Endo-Free plasmid kits
Supplier:  OMEGA BIO-TEK
Certificates
E.Z.N.A.® Endo-Free plasmid kits
Supplier:  OMEGA BIO-TEK
Certificates

Specifications

  • Method / Format:
    Column (silica-based)
  • Prep size:
    Midi
  • Refine target molecule:
    Plasmid (endotoxin-free)
  • Sample type:
    Bacteria
  • Target molecule:
    Plasmid
  • Sample size:
    15 - 50 ml
  • Description:
    E.Z.N.A.® Endo-Free Plasmid Midi Kit

Specifications

Related Information

Frequently Bought Together