Order Entry
Austria
ContactUsLinkComponent
E.Z.N.A.® and E-Z 96® Fastfilter® Plasmid purification kits
E.Z.N.A.® and E-Z 96® Fastfilter® Plasmid purification kits
Supplier:  OMEGA BIO-TEK
Certificates
E.Z.N.A.® and E-Z 96® Fastfilter® Plasmid purification kits
Supplier:  OMEGA BIO-TEK

Specifications

  • Method / Format:
    Multiwell plate
  • Prep size:
    Mini
  • Refine target molecule:
    Plasmid
  • Sample type:
    Bacteria
  • Target molecule:
    Plasmid
  • Sample size:
    700 μl - 1 ml/well
  • Description:
    E-Z 96® Fastfilter® Plasmid Kit

Specifications

Frequently Bought Together