ContactUsLinkComponent
Replacement plunger, for blue line autosampler
Replacement plunger, for blue line autosampler
  HEWLG4513-80227
 :  Agilent
undefined
Replacement plunger, for blue line autosampler
  HEWLG4513-80227
 :  Agilent
  • For:
    10 µl Blue Line autosampler syringes
  • Description:
    Replacement plunger for fixed needle
  • Seq:
    12
  • Pk:
    1
PTFE tip plunger for blue line autosampler fixed needle syringes for 7693A ALS.