Unions
Supplier: VICI JOUR RESEARCH
Jump to:
About this item
Unions for easy connection of low pressure solvent lines.
- Low dead volume
- Inert and biocompatible
7 Options Available Below
Product Details & Documents
Unions for easy connection of low pressure solvent lines.
- Low dead volume
- Inert and biocompatible
Product Family Options
Polyamide
Product Information
- MaterialAvailabilityPrice
- JOURJR-060-5Union, through bore LP body only, 1/4”-28, for flanged tubingUnion, through bore LP body only, 1/4”-28, for flanged tubingPolyamideLog in for availability1 * 5 ST29,10 €
PEEK
Product Information
- MaterialAvailabilityPrice
- JOURJR-064Union, LP 0,5 mm bore, body only
- JOURJR-068Union, LP 0,75 mm bore, body only
- JOURJR-065Union, LP 1,3 mm bore, body only
- 554-1026Union, LP 0,5 mm bore, complete, 1/16” tubing
- JOURJR-1068Union, LP 0,75 mm bore, complete, 1/16” tubing
- 554-1027Union, LP 1,3 mm bore, complete, 1/8” tubing