Reagent containers for Microlab® STAR line
Supplier: HAMILTON ROBOTICS
Certificates
About this item
PP, white or black. These reagent reservoirs with 60, 120 or 200 ml capacity can be placed in a reagent carrier on the STAR line instruments. The reservoirs are available self-standing or non-self-standing.
- Special bottom geometry minimises residual volumes
- With volume markings, without or with lid
- With one (60 ml), three (120 ml) or four (200 ml) wave breakers to avoid splashes during movements of the filled containers
- IVD compliant
- Autoclavable
Ordering information: The carriers are not included and need to be ordered separately.
Frequently Bought Together