Septa for 8 mm screw top vials
: Agilent
Frequently Bought Together
Ethanol 70% ≥70% (v/v), TechniSolv®
: VWR Chemicals
Hypodermic needles, HENKE-JECT® for single use
: HENKE SASS WOLF
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR®, Short Thread Vials ND9
: VWR Collection
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
VWR®, Screw Caps for Short Thread Vials ND9
: VWR Collection
VWR®, Snap Cap Vials
: VWR Collection
VWR®, Crimp Neck Vials ND11
: VWR Collection