Order Entry
Austria
Orders LinkContactUsLinkComponent
Storage solution for ROSS electrodes

Storage solution for ROSS electrodes

ROSS® electrode storage solution (475 ml bottle), 475 mL
Catalog # 662-1548
Supplier:   Thermo Orion
Storage solution for ROSS electrodes
Catalog # 662-1548
Supplier:  Thermo Orion
CAS Number:  
Storage solution for ROSS electrodes

ROSS® electrode storage solution (475 ml bottle), 475 mL

CAS Number:  
Price
92,60 €
Log in for availability
1 * 475 mL
Product Details & Documents

Restore proper electrode performance and prolong electrode life using Thermo Scientific™ Orion™ pH electrode storage solutions.

  • Storage solution ensures that the pH electrode works properly after prolonged storage, overnight storage and between samples

 

Specifications

Specifications

  • Pk:
    475 mL
  • Description:
    ROSS® electrode storage solution (475 ml bottle)

Specifications