ContactUsLinkComponent
Hepatocyte culture medium, Corning®
Hepatocyte culture medium, Corning®
Catalog # 734-2386
Supplier:  Corning
   Low Manufacturing Impact
CAS Number:  
undefined
Hepatocyte culture medium, Corning®
Catalog # 734-2386
Supplier:  Corning
   Low Manufacturing Impact
CAS Number:  
Specifications
  • Pk:
    500 mL
  • Description:
    Hepatocyte Culture Medium Kit
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    ISO 14001 - Bedford
Specifications
About this item
Hepatocyte culture medium is a serum-free, fully-defined medium optimised for hepatocyte differentiation.
Primary human hepatocytes are fresh-couriered in exclusive Falcon® Flip-Lock packaging to ensure best performance and viability upon arrival - without irritating film membranes, gel stabilizers or preservatives that can interfere with your research. Excellent viability- shipped in Corning Hepato-STIM hepatocyte culture medium on Corning® Biocoat™ Multiwell Plates and Flasks for maximal performance. Rapid response- As soon as we have donor tissue, you will be notified instantly as per your instructions.
Delivery: Kit comprises hepatocyte culture medium (500 ml) and EGF (5 µg).