Orders LinkContactUsLinkComponent
Handheld refractometer, IFT
Handheld refractometer, IFT
Industrial fluid tester, IFT 40, 0 - 30, 1,3330 - 1,3860 nD, 0,5
Catalog # 635-2110
Supplier:   REICHERT
Handheld refractometer, IFT
Catalog # 635-2110
Supplier:  REICHERT
Handheld refractometer, IFT
Industrial fluid tester, IFT 40, 0 - 30, 1,3330 - 1,3860 nD, 0,5
Price
352,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
This handheld refractometer is designed to determine ratio or concentration of water-soluble industrial fluids or lubricants.
Specifications
  • Model:
    IFT 40
  • Range:
    0 - 30
  • Resolution:
    0,5
  • Description:
    Industrial fluid tester
  • Seq:
    02
  • Refractive index:
    1,3330 - 1,3860 nD
  • Cat. No.:
    635-2110
  • Scales:
    0,5