PCR plates, 96-well, elevated skirt, Axygen®
Supplier: Corning
Certificates
About this item
PP plates, semi-skirted. Axygen® 96-well elevated skirt PCR plate is designed to fit ABI Real-Time PCR thermal cyclers and sequencers. Can be sealed with 8/12 strip caps, AxyMat™ or sealing film.
- Ultra-thin walls of uniform thickness accelerate heat transfer, leading to shorter cycle times and more consistent results
- Certified free from RNase, DNase and non pyrogenic for highest sample integrity
- Single notch corner
- AxyMat™ sealing mats autoclavable and reusable
Specifications
- Catalog No:
- 732-0665
- 731-0692
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa