Orders LinkContactUsLinkComponent
Stackable drum sumps and pallets, WSP
Stackable drum sumps and pallets, WSP
Stackable drum sumps and pallets, WSP
Certificates
Stackable drum sumps and pallets, WSP
10 Options Available Below
Product Details & Documents
These stackable drum sumps and pallets are made of stable construction from 3 mm sheet steel.
  • Sump capacity of 220 L
  • Ground clearance 100 mm for internal transport
  • Space-savings as models WSP-STS and SKS can be double-stacked
  • Corrosion protection by primer coating and painting or fire-galvanisation
Product Family Options
Fire-galvanised
Product Information
  • W×D×H
    Availability
    Price
  • LACOP21-1451-A
    Sump pallets, with capacity 2×200 l drums, WSP-2SSS
    Sump pallets, with capacity 2×200 l drums, WSP-2SSS
    1410×810×1300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • LACOP21-1411-A
    Sump pallets, with capacity 2×200 l drums, WSP-2STS
    Sump pallets, with capacity 2×200 l drums, WSP-2STS
    1410×810×1400 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • LACOP21-1421-A
    Sump pallets, with capacity 2×200 l drums, WSP-2SKS
    Sump pallets, with capacity 2×200 l drums, WSP-2SKS
    1410×810×1400 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • LACOP21-1412-A
    Sump pallets, with capacity 4×200 l drums, WSP-4STS
    Sump pallets, with capacity 4×200 l drums, WSP-4STS
    1410×1290×1320 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • LACOP21-1422-A
    Sump pallets, with capacity 4×200 l drums, WSP-4SKS
    Sump pallets, with capacity 4×200 l drums, WSP-4SKS
    1410×1290×1320 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
Painted
Product Information
  • W×D×H
    Availability
    Price
  • LACOP21-2451-A
    Sump pallets, with capacity 2×200 l drums, WSP-2SSS
    Sump pallets, with capacity 2×200 l drums, WSP-2SSS
    1410×810×1300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • LACOP21-2411-A
    Sump pallets, with capacity 2×200 l drums, WSP-2STS
    Sump pallets, with capacity 2×200 l drums, WSP-2STS
    1410×810×1400 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • LACOP21-2421-A
    Sump pallets, with capacity 2×200 l drums, WSP-2SKS
    Sump pallets, with capacity 2×200 l drums, WSP-2SKS
    1410×810×1400 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • LACOP21-2412-A
    Sump pallets, with capacity 4×200 l drums, WSP-4STS
    Sump pallets, with capacity 4×200 l drums, WSP-4STS
    1410×1290×1320 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • LACOP21-2422-A
    Sump pallets, with capacity 4×200 l drums, WSP-4SKS
    Sump pallets, with capacity 4×200 l drums, WSP-4SKS
    1410×1290×1320 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Tension belt
    Tension belt
    LACOZ50-1920-A
    Log in for availability
    1 * 1 ST
Product Information
  • Catalog #
    Availability
    Price
  • Safety chain
    Safety chain
    LACOP21-2470-A
    Log in for availability
    1 * 1 ST