Order Entry
Austria
ContactUsLinkComponent
Eosin Y (yellowish) in aqueous solution, Q Path® for microscopy
Eosin Y (yellowish) in aqueous solution, Q Path® for microscopy
Supplier:  VWR Chemicals
Certificates
undefined
Eosin Y (yellowish) in aqueous solution, Q Path® for microscopy
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Appearance:
    Clear liquid
  • Colour of solution:
    Orange-red
  • Density (20/4):
    0.9 - 1.1
  • pH (20°C):
    5 - 5.5
  • Absorbance (0.1 %):
    514 - 518 nm

Specification Test Results

Frequently Bought Together