Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Furniture
Storage Cabinets
Accessories for storage cabinets for acids and alkalis SLS 600
Accessories for storage cabinets for acids and alkalis SLS 600
Accessories for storage cabinets for acids and alkalis SLS 600
Catalog # LACOB80-2824-A
Supplier:
LACONT UMWELTTECHNIK
Expand image
Accessories for storage cabinets for acids and alkalis SLS 600
Catalog # LACOB80-2824-A
Supplier:
LACONT UMWELTTECHNIK
Specifications
Description:
Shelves, pull-out
Pk:
1
For:
SLS 600
Specifications
About this item
Product Details
Product Details