Orders LinkContactUsLinkComponent
Tubes and systems for active sampling
Tubes and systems for active sampling
Supplier:   Dräger
Tubes and systems for active sampling
Supplier:  Dräger
Certificates
Tubes and systems for active sampling
Tubes and systems for active sampling
16 Options Available Below
Product Details & Documents
Sampling activated tubes for direct measurement indication even with complex compounds and mixtures of substances in air investigations and gas analyses.
  • High precision guaranteed
  • Very reliable in use
  • Improved colour depth and contrast using an optimised production method
Product Family Options
Product Information
  • Type
    Availability
    Price
  • DRAE6400741
    Activated charcoal tubes
    Activated charcoal tubes
    NIOSH
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 227,00 €
    227,00 €
    1 * 50 ST
       227,00 €
  • 151-3146
    Activated charcoal tubes
    Activated charcoal tubes
    BIA
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 59,00 €
    59,00 €
    1 * 10 ST
       59,00 €
  • DRAE6400743
    Activated charcoal tubes
    Activated charcoal tubes
    BIA
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 243,00 €
    243,00 €
    1 * 50 ST
       243,00 €
  • DRAE6400744
    Activated charcoal tube
    Activated charcoal tube
    BIA
    Log in for availability
    Log in for availability
    1 * 200 ST
    1 * 200 ST - 830,00 €
    830,00 €
    1 * 200 ST
       830,00 €
  • DRAE6728831
    Activated charcoal tubes
    Activated charcoal tubes
    G
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 59,00 €
    59,00 €
    1 * 10 ST
       59,00 €
  • DRAE6400745
    Activated charcoal tubes
    Activated charcoal tubes
    G
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 243,00 €
    243,00 €
    1 * 50 ST
       243,00 €
  • DRAE6400746
    Activated charcoal tubes
    Activated charcoal tubes
    G
    Log in for availability
    Log in for availability
    1 * 200 ST
    1 * 200 ST - 830,00 €
    830,00 €
    1 * 200 ST
       830,00 €
  • DRAE8101821
    Activated charcoal tubes
    Activated charcoal tubes
    B/G
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 59,00 €
    59,00 €
    1 * 10 ST
       59,00 €
  • DRAE6728891
    Diffusion samplers
    Diffusion samplers
    ORSA 5
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 125,00 €
    125,00 €
    1 * 5 ST
       125,00 €
  • DRAE6728919
    Diffusion samplers
    Diffusion samplers
    ORSA 25
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 459,00 €
    459,00 €
    1 * 25 ST
       459,00 €
  • DRAE6400131
    Isocyanate sampling set and analyse
    Isocyanate sampling set and analyse
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 274,00 €
    274,00 €
    1 * 1 ST
       274,00 €
  • DRAE6400271
    Aldehyde sampling set and analyse
    Aldehyde sampling set and analyse
    Log in for availability
    Log in for availability
    1 * 1 SET
    1 * 1 SET - 274,00 €
    274,00 €
    1 * 1 SET
       274,00 €
  • DRAE8101271
    Sampling tubes amines
    Sampling tubes amines
    TEA
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 104,00 €
    104,00 €
    1 * 10 ST
       104,00 €
  • 151-3145
    Silica gel tubes
    Silica gel tubes
    BIA
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 57,25 €
    57,25 €
    1 * 10 ST
       57,25 €
  • DRAE6728851
    Silica gel tubes
    Silica gel tubes
    G
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 57,25 €
    57,25 €
    1 * 10 ST
       57,25 €
  • DRAE8101861
    Silica gel tubes
    Silica gel tubes
    B/G
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 57,25 €
    57,25 €
    1 * 10 ST
       57,25 €