0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Austria
ContactUsLinkComponent
Standards
Environmental Analysis Standards
VWR®, Hydrazine standard solutions in water
VWR®, Hydrazine standard solutions in water
Catalog # 87344.180
Supplier:
VWR Chemicals
CAS Number:
SDS
Certificates
Expand image
VWR®, Hydrazine standard solutions in water
Catalog # 87344.180
Supplier:
VWR Chemicals
CAS Number:
SDS
Certificates
Specifications
Concentration:
100 mg/l
Pk:
100 mL
Description:
Standard solution hydrazine
Specifications
About this item
Product Details
Documentation
Product Details
Documentation