Order Entry
Austria
ContactUsLinkComponent
Ethanol, Q Path® fixative reagent, for microscopy
Flammable
Ethanol, Q Path® fixative reagent, for microscopy
Catalog # ALC0060AF59001
Supplier:  VWR Chemicals
CAS Number:  
undefined
Flammable
Ethanol, Q Path® fixative reagent, for microscopy
Catalog # ALC0060AF59001
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Pot
  • Pk:
    50 x 20 ml
  • Seq:
    0050
  • Pk Info:
    60 ml pots contain 20ml

Specifications

Specification Test Results

  • Appearance:
    Colourless liquid
  • Density (20/4):
    0,922 - 0,935

Specification Test Results

About this item

 :
 :46.07 g/mol
Storage Temperature:Ambient
EINECS:200-578-6
UN:1170
ADR:3,III
 :GHS02
 :1
CAS Number:64-17-5
Formula:C₂H₅OH
MDL Number:MFCD00003568