Order Entry
Austria
ContactUsLinkComponent
Sodium thiosulphate, anhydrous ≥98%, GPR RECTAPUR®
Sodium thiosulphate, anhydrous ≥98%, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates
undefined
Sodium thiosulphate, anhydrous ≥98%, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay:
    Min. 98 %
  • pH (20°C; 5 %):
    6.0 - 8.5
  • S (Sulphide):
    Max. 50 ppm
  • Ca (Calcium):
    Max. 50 ppm
  • Cd (Cadmium):
    Max. 50 ppm
  • Co (Cobalt):
    Max. 50 ppm
  • Cu (Copper):
    Max. 50 ppm
  • Fe (Iron):
    Max. 50 ppm
  • K (Potassium):
    Max. 200 ppm
  • Ni (Nickel):
    Max. 50 ppm
  • Pb (Lead):
    Max. 50 ppm
  • Zn (Zinc):
    Max. 50 ppm

Specification Test Results

Frequently Bought Together