Order Entry
Austria
ContactUsLinkComponent
Methyl red solution in ethanol acid-base indicator, Fluka™
Irritant
Flammable
Methyl red solution in ethanol acid-base indicator, Fluka™
Catalog # 32941-100ML
CAS Number:  
Irritant
Flammable
Methyl red solution in ethanol acid-base indicator, Fluka™
Catalog # 32941-100ML
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

Specification Test Results

  • Change:
    complying
  • Test of sensitivity:
    complying

Specification Test Results

About this item

 :
Flash Pt:28 °C
EINECS:207-776-1
ADR:3,III
 :269.3 g/mol
UN:1170
 :GHS07
 :GHS02
Storage Temperature:Ambient
CAS Number:493-52-7
Formula:C₁₅H₁₅N₃O₂
MDL Number:MFCD00002425