Order Entry
Austria
ContactUsLinkComponent
Methyl red 0.1 g/l in ethanol 57% (v/v), Millipore®
Flammable
Methyl red 0.1 g/l in ethanol 57% (v/v), Millipore®
Catalog # SIAL08714-100ML-F
Supplier:  Merck
CAS Number:  
Flammable
Methyl red 0.1 g/l in ethanol 57% (v/v), Millipore®
Catalog # SIAL08714-100ML-F
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

About this item

 :
ADR:3,II
 :M-Clarity™ quality level = MQ100
 :269.3 g/mol
EINECS:207-776-1
UN:1170
 :GHS02
Storage Temperature:Ambient
CAS Number:493-52-7
Formula:C₁₅H₁₅N₃O₂
MDL Number:MFCD00002425