Buffers, sodium borate, 0,5 M solutions
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS No.: 1330-43-4
MDL No.: MFCD00081185
Specifications
- CAS No.:1330-43-4
- MDL No.:MFCD00081185
- Physical form:Liquid
- Appearance:Colourless
Specifications
Related Information
Frequently Bought Together