Buffers, MES, 0,5 M solutions
CAS No.: 145224-94-8
Physical Form: Liquid
Appearance: Colourless
MDL No.: MFCD00283848
- CAS No.:145224-94-8
- MDL No.:MFCD00283848
- Physical form:Liquid
- Appearance:Colourless
Frequently Bought Together
CAS No.: 145224-94-8
Physical Form: Liquid
Appearance: Colourless
MDL No.: MFCD00283848