Digital Solutions
Applications
Support
About
Austria
ContactUsLinkComponent
Guard Column Holders
Guard cartridge holder, HyperREZ™ XP
Guard cartridge holder, HyperREZ™ XP
HYPE60002-354
:
Thermo Fisher Scientific
Guard cartridge holder, HyperREZ™ XP
HYPE60002-354
:
Thermo Fisher Scientific
Description:
Guard cartridge holder, for HyperREZ™ XP 3×5 mm guard cartridges
Pk:
1