Incinerating dishes, rectagular
Supplier: MORGAN ADVANCED MATERIALS HALD
Jump to:
About this item
Alsint 99,7.
- Good thermal shock resistance due to high thermal conductivity
- Resistant to chemical attack, e.g. hydrogen and other reducing gases
12 Options Available Below
Product Details & Documents
Alsint 99,7.
- Good thermal shock resistance due to high thermal conductivity
- Resistant to chemical attack, e.g. hydrogen and other reducing gases
Product Family Options
Product Information
- LengthHeightWidthAvailabilityPrice