Order Entry
Austria
ContactUsLinkComponent
Diode laser, green, Class II and IIIa
Diode laser, green, Class II and IIIa
Catalog # DIDAPOD013132
Supplier:  DIDALAB
undefined
Diode laser, green, Class II and IIIa
Catalog # DIDAPOD013132
Supplier:  DIDALAB

Specifications

  • Description:
    Diode laser, green, Class II and IIIa
  • Pk:
    1
  • Wavelength:
    532 nm

Specifications

About this item

Extremely luminous beam, ideal for classroom experiments. Class IIIa (5 mW Power)

  • Highly miniaturised system with glass optic components
Delivery: Provided with Ø 10 mm rod, mains power supply & adapter for use in Class II.