About this item
Carcass construction, i.e. each cabinet can stand alone. Carcass glued.
Material: Melamine resin coated chipboard in accordance with DIN 68765, emission class E1, all front edges covered with 2 mm ABS, rear edge of the carcass with 0,6 mm ABS. Sides and fronts 19 mm thick, base, panels and shelves 22 mm thick, rear panel 8 mm thick.
Panels/base: 22 mm thick, with 2 mm ABS edging on the front. Height adjustable in 32 mm increments with recessed shelf supports. Sides and fronts 19 mm thick.
Height adjustment: With four plastic guides, 20 mm high, 80 mm diameter, adjustable from inside through the base of the cabinet (+10 mm).
Steel base (optional): 7 cm high, cabinet can be made 5,1 cm higher with the steel base than with the adjustable feet.
Doors: 125-degree opening angle with sealing lip.
Lock: 3-point locking mechanism with door damper buffer.
Hinges: High quality, all metal cup hinges manufactured by Hettich. Quick mounting technology, 3-dimensionally adjustable.