Nuts and ferrules, Super Flangeless™, Upchurch Scientific®
Supplier: UPCHURCH SCIENTIFIC
Certificates
About this item
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
Accessories for Mettler balances
Supplier: Mettler - Toledo
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
Wipes for delicate tasks, Kimtech™ Science
Supplier: KIMBERLY CLARK
VWR® Facial Tissues
Supplier: VWR Collection
VWR®, Nitrile Gloves
Supplier: VWR Collection
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
VWR®, Glove Box Holders
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
pH electrodes, SenTix® PLUS
Supplier: WTW
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Plastic cuvette for LICO
Supplier: Hach
Reversed phase chromatography columns, RSpak DS
Supplier: Resonac