Order Entry
Austria
ContactUsLinkComponent
Sodium hydroxide 0.1 mol/l (0.1 N) in aqueous solution, Supelco®
Sodium hydroxide 0.1 mol/l (0.1 N) in aqueous solution, Supelco®
Catalog # 43617-1L
Supplier:  Merck
CAS Number:  
undefined
Sodium hydroxide 0.1 mol/l (0.1 N) in aqueous solution, Supelco®
Catalog # 43617-1L
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :40 g/mol
Density:1 g/cm³ (25 °C)
Storage Temperature:Ambient
UN:1824
ADR:8,III
 :M-Clarity™ quality level = MQ100
EINECS:215-185-5
CAS Number:1310-73-2
Formula:NaOH
MDL Number:MFCD00003548