Order Entry
Austria
ContactUsLinkComponent
Potassium chloride 0.001 mol/l (0.001 N) in aqueous solution
Potassium chloride 0.001 mol/l (0.001 N) in aqueous solution
Catalog # HACHC20C280
Supplier:  Hach
CAS Number:  
Potassium chloride 0.001 mol/l (0.001 N) in aqueous solution
Catalog # HACHC20C280
Supplier:  Hach
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    500 mL
  • Seq:
    2725

Specifications

About this item

 :
 :74.55 g/mol
Storage Temperature:Ambient
EINECS:231-211-8
CAS Number:7447-40-7
Formula:KCl
MDL Number:MFCD00011360