Order Entry
Austria
ContactUsLinkComponent
Sodium thiosulphate pentahydrate 99.5-101.0% (by iodometry), crystals, BAKER ANALYZED® ACS, J.T.Baker®
Sodium thiosulphate pentahydrate 99.5-101.0% (by iodometry), crystals, BAKER ANALYZED® ACS, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
Sodium thiosulphate pentahydrate 99.5-101.0% (by iodometry), crystals, BAKER ANALYZED® ACS, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • Meets ACS Reagent Chemical Requirements:
  • For Laboratory,Research,or Manufacturing Use:
    99.5 - 101.0 %
  • Assay (Na₂S₂O₃·5H₂O) (by iodometry):
    99.5 - 101.0 %
  • Insoluble Matter:
    ≤0.005 %
  • pH of 5% Solution at 25°C:
    6.0 - 8.4
  • Nitrogen Compounds (as N):
    ≤0.002 %
  • Sulfate and Sulfite (as SO₄):
    ≤0.1 %
  • Sulfide (S):
    Passes Test

Specification Test Results

Related Information

Frequently Bought Together