Order Entry
Austria
ContactUsLinkComponent
Potassium chloride 3 mol/l (3 N) in aqueous solution
Potassium chloride 3 mol/l (3 N) in aqueous solution
Catalog # WTWA109706
Supplier:  WTW
CAS Number:  
Potassium chloride 3 mol/l (3 N) in aqueous solution
Catalog # WTWA109706
Supplier:  WTW
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    50 mL
  • Seq:
    2370
  • Environmentally Preferable:

Specifications

About this item

Synonyms: Potassiumchloride
 :
 :74.55 g/mol
Density:1.13 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-211-8
CAS Number:7447-40-7
Formula:KCl
MDL Number:MFCD00011360