Order Entry
Austria
ContactUsLinkComponent
Sodium thiosulphate 20 g/L in aqueous solution, Sigma-Aldrich®
Sodium thiosulphate 20 g/L in aqueous solution, Sigma-Aldrich®
Catalog # HT1005-100ML
CAS Number:  
undefined
Sodium thiosulphate 20 g/L in aqueous solution, Sigma-Aldrich®
Catalog # HT1005-100ML
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

Specification Test Results

  • Appearance (Colour):
    Colourless
  • Appearance (Form):
    Solution
  • Appearance (Turbidity), Essentially free of particles/precipitate:
    Clear
  • Performance:
    Conforms
  • Expiration Date Period:
    24 months

Specification Test Results

About this item

 :
 :158.11 g/mol
Density:1010 g/cm³
Storage Temperature:Refrigerator
EINECS:231-867-5
CAS Number:7772-98-7
Formula:Na₂S₂O₃
MDL Number:MFCD00003499