
Nitric acid (1:1) (v/v) 38% (v/v)
Catalog # 705-1124
Supplier: Hach
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:500 mL
- Seq:2725
Specifications
About this item
:
:63.01 g/mol
Boiling Pt:∼86 °C (1013 hPa)
Melting Pt:–42 °C
Density:1.2…1.4 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-714-2
UN:2031
ADR:8,II
:GHS05
CAS Number:7697-37-2
Formula:HNO₃
MDL Number:MFCD00011349