Order Entry
Austria
ContactUsLinkComponent
Potassium dihydrogen phosphate 1 mol/l in aqueous solution
Potassium dihydrogen phosphate 1 mol/l in aqueous solution
Catalog # 786-488
Supplier:  G-Biosciences
CAS Number:  
Potassium dihydrogen phosphate 1 mol/l in aqueous solution
Catalog # 786-488
Supplier:  G-Biosciences
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :136.09 g/mol
Storage Temperature:Ambient
EINECS:231-913-4
CAS Number:7778-77-0
Formula:KH₂PO₄
MDL Number:MFCD00011401