Conductivity standards, potassium chloride, referenced at 25 °C
Supplier: Hach
Certificates
Certificates
About this item
Frequently Bought Together
Membrane filters, Microsart® e.motion
Supplier: Sartorius
Calibration standards for Turbiquant®, Supelco®
Supplier: Merck
pH buffers, DuraCal
Supplier: HAMILTON BONADUZ
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
Sulphuric acid 2 mol/l (4 N) in aqueous solution, AVS TITRINORM volumetric solution
Supplier: VWR Chemicals
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Lens cleaning tissues, Grade 105, Whatman™
Supplier: Whatman products (Cytiva)
VWR®, Nitrile Gloves
Supplier: VWR Collection
VWR® Culture Media, Ready To Use, In Plates
Supplier: VWR Chemicals