Order Entry
Austria
ContactUsLinkComponent
Potassium hydroxide 45% (w/v) in aqueous solution
Potassium hydroxide 45% (w/v) in aqueous solution
Catalog # 47010.K2
CAS Number:  
Potassium hydroxide 45% (w/v) in aqueous solution
Catalog # 47010.K2
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :56.11 g/mol
Density:1.44
Storage Temperature:Ambient
EINECS:215-181-3
UN:1814
ADR:8,II
 :GHS05
 :GHS07
CAS Number:1310-58-3
Formula:KOH
MDL Number:MFCD00003553