Buffer, TAE solution 50X concentrate (TRIS-acetate-EDTA buffer)
Physical Form: Liquid
Appearance: Colourless
MDL No.: MFCD00236357
- MDL No.:MFCD00236357
- Physical form:Liquid
- Appearance:Colourless
Frequently Bought Together
Physical Form: Liquid
Appearance: Colourless
MDL No.: MFCD00236357